3-Amino-5-bromo-2-methoxypyridine 96%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
5-Bromo-2-methoxypyridin-3-amine
H54299.03EA
306
EUR
H54299.03
H54299.06
H54299.MD
3-Amino-5-bromo-2-methoxypyridine 96%
3-Amino-5-bromo-2-methoxypyridine
Formula:
C₆H₇BrN₂O MW: 203,04 g/mol Melting Pt: 53…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD07368887 CAS Number: 884495-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...