1,2-Vinylenebis(triphenylphosphonium bromide) monohydrate ≥98%
Supplier: Thermo Scientific
|
Warning
|
L00956.09EA
41.3
EUR
L00956.09
1,2-Vinylenebis(triphenylphosphonium bromide) monohydrate ≥98%
1,2-Vinylenebis(triphenylphosphonium bromide) monohydrate
Formula:
C₃₈H₃₂Br₂P₂·1H₂O MW: 728,45 g/mol (710,44 anhy) Melting Pt: ∼ 236…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040548 CAS Number: 54770-27-3 EINECS: 259-334-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...