Ethyl dibromofluoroacetate ≥97%
Supplier: Thermo Scientific
|
Danger
|
L16491.14EA
240
EUR
L16491.14
L16491.06
Ethyl dibromofluoroacetate ≥97%
Ethyl dibromofluoroacetate
Formula:
C₄H₅Br₂FO₂ MW: 263,89 g/mol Boiling Pt: 82 °C (33 mmHg) Density: 1,894 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042068 CAS Number: 565-53-7 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...