Linalyl acetate (synthetic) 95%
Supplier: Thermo Scientific
|
Warning
|
ACRO232305000EA
78
EUR
ACRO232305000
ACRO232300050
ACRO232301000
Linalyl acetate (synthetic) 95%
Linalyl acetate
Formula:
C₁₂H₂₀O₂ Flash Pt: 85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008907 CAS Number: 115-95-7 EINECS: 204-116-4 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4490 to 1.4520 (20°C, 589 nm) |
GC | ≥94 % |
Specific gravity | (20°C) 0.898 to 0.903 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...