4-Pentenyl acetate ≥98%
Supplier: Thermo Scientific
|
Warning
|
B20258.14EA
249
EUR
B20258.14
B20258.06
4-Pentenyl acetate ≥98%
4-Pentenyl acetate
Formula:
C₇H₁₂O₂ MW: 128,17 g/mol Boiling Pt: 144…146 °C Density: 0,91 Flash Pt: 46 °C (115 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008723 CAS Number: 1576-85-8 EINECS: 216-413-6 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...