(2,6-Dichlorobenzyl)triphenylphosphonium chloride ≥98%

Supplier: Thermo Scientific
Warning

A17651.06EA 25.6 EUR
A17651.06 A17651.14 A17651.22
(2,6-Dichlorobenzyl)triphenylphosphonium chloride ≥98%
(2,6-Dichlorobenzyl)triphenylphosphonium chloride
Formula: C₂₅H₂₀Cl₃P
Storage Temperature: Ambient
MDL Number: MFCD00061392
CAS Number: 18583-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR