4-Hydroxy-2,6-dimethoxybenzaldehyde
Supplier: Molekula
Synonyms:
2,6-Dimethoxy-4-hydroxybenzaldehyde
MOLEM42566154EA
0
EUR
MOLEM42566154
4-Hydroxy-2,6-dimethoxybenzaldehyde
4-Hydroxy-2,6-dimethoxybenzaldehyde
Formula:
C₉H₁₀O₄ MW: 182,1733 g/mol |
MDL Number:
MFCD01407645 CAS Number: 22080-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...