(1S)-(+)-Camphor-10-sulphonamide ≥97%
Supplier: Thermo Scientific
44361.03EA
52.3
EUR
44361.03
44361.06
(1S)-(+)-Camphor-10-sulphonamide ≥97%
(1S)-(+)-Camphor-10-sulphonamide
Formula:
C₁₀H₁₇NO₃S MW: 231,32 g/mol Melting Pt: 133…135 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00075611 CAS Number: 60933-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...