7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid hydrate 95%

Supplier: Thermo Scientific
Warning

H35003.MD
H35003.MDEA 171 EUR
H35003.MD
7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid hydrate 95%
7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Formula: C₈H₅BrN₂O₂·H₂O
MW: 241,05 g/mol (anhy)
Melting Pt: 234…235 °C
Storage Temperature: Ambient
MDL Number: MFCD22989340
CAS Number: 1019018-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR