2-Fluoro-1-methylpyridinium-p-toluenesulphonate ≥90%, tech.

Warning

Synonyms: 2-Fluoro-1-methylpyridin-1-ium 4-methylbenzenesulphonate

L11088.06EA 89.3 EUR
L11088.06 L11088.03
2-Fluoro-1-methylpyridinium-p-toluenesulphonate ≥90%, tech.
2-Fluoro-1-methylpyridinium p-toluenesulphonate
Formula: C₁₃H₁₄FNO₃S
MW: 283,32 g/mol
Melting Pt: 128…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00011983
CAS Number: 58086-67-2
EINECS: 261-108-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR