2-Amino-5,6-dimethylbenzimidazole, Sigma-Aldrich®
Supplier: SIGMA-ALDRICH MICROSCOPY
A51203-25G
A51203-5G
A51203-25GEA
756
EUR
A51203-25G
A51203-5G
2-Amino-5,6-dimethylbenzimidazole, Sigma-Aldrich®
2-Amino-5,6-dimethylbenzimidazole
Formula:
C₉H₁₁N₃ |
MDL Number:
MFCD00005597 CAS Number: 29096-75-1 EINECS: 249-430-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...