1-Acetyl-3-thiosemicarbazide ≥97%

Supplier: Thermo Scientific

Synonyms: 2-Acetylhydrazinecarbothioamide

A16915.22EA 0 EUR
A16915.22
1-Acetyl-3-thiosemicarbazide ≥97%
1-Acetyl-3-thiosemicarbazide
Formula: C₃H₇N₃OS
Melting Pt: 165…168 °C
MDL Number: MFCD00004932
CAS Number: 2302-88-7
EINECS: 218-956-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR