Methyl-4-methyl-3-nitrobenzoate ≥99%
Supplier: Thermo Scientific
Synonyms:
Methyl 4-methyl-3-nitrobenzoate
B25299.22EA
592
EUR
B25299.22
Methyl-4-methyl-3-nitrobenzoate ≥99%
Methyl 4-methyl-3-nitrobenzoate
Formula:
C₉H₉NO₄ MW: 195,18 g/mol Melting Pt: 49…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00130054 CAS Number: 7356-11-8 EINECS: 230-886-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...