2-Chloro-5-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: Thermo Scientific
Warning

B23937.06EA 118 EUR
B23937.06 B23937.03
2-Chloro-5-(trifluoromethyl)benzyl alcohol ≥97%
2-Chloro-5-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆ClF₃O
MW: 210,58
Melting Pt: 57…60 °C
Storage Temperature: Ambient
MDL Number: MFCD01631469
CAS Number: 64372-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR