2-Methoxy-6-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: Thermo Scientific
Warning

H32539.MDEA 38.2 EUR
H32539.MD H32539.03
2-Methoxy-6-(trifluoromethyl)benzyl alcohol ≥97%
2-Methoxy-6-(trifluoromethyl)benzyl alcohol
Formula: C₉H₉F₃O₂
Melting Pt: 73 °C…75 °C
Storage Temperature: Ambient
MDL Number: MFCD09832325
CAS Number: 1017779-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR