4-(Benzyloxy)phenyl isothiocyanate ≥98%
Supplier: Thermo Scientific
|
Danger
|
L10038.06EA
238
EUR
L10038.06
4-(Benzyloxy)phenyl isothiocyanate ≥98%
4-(Benzyloxy)phenyl isothiocyanate
Formula:
C₁₄H₁₁NOS MW: 241,31 g/mol Melting Pt: 62…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041095 CAS Number: 139768-71-1 EINECS: 000-000-0 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...