(S)-(+)-2-Heptyl isothiocyanate ≥98%

Supplier: Thermo Scientific
Danger

L20065.03EA 91.6 EUR
L20065.03
(S)-(+)-2-Heptyl isothiocyanate ≥98%
(S)-(+)-2-Heptyl isothiocyanate
Formula: C₈H₁₅NS
Density: 0,910
MDL Number: MFCD05664052
CAS Number: 737000-94-1
EINECS: 000-000-0
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR