(S)-(+)-2-Hexyl isothiocyanate 95%
Supplier: Thermo Scientific
|
Danger
|
L20067.03EA
96.3
EUR
L20067.03
(S)-(+)-2-Hexyl isothiocyanate 95%
(S)-(+)-2-Hexyl isothiocyanate
Formula:
C₇H₁₃NS MW: 143,25 g/mol Density: 0,91 |
MDL Number:
MFCD05664054 CAS Number: 737000-96-3 EINECS: 000-000-0 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...