3'-Isothiocyanatoacetophenone ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 3-Acetylphenyl isothiocyanate

L10134.06EA 82.1 EUR
L10134.06 L10134.03
3'-Isothiocyanatoacetophenone ≥97%
3'-Isothiocyanatoacetophenone
Formula: C₉H₇NOS
Boiling Pt: 135…136 °C (2 mmHg)
Melting Pt: ∼ 24 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041081
CAS Number: 3125-71-1
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR