Charcoal activated 0.3-0.5 mm (35-50 mesh) for gas chromatography, Supelco®

Synonyms: Carbon activated,Activated carbon,Activated charcoal

M-Clarity™ quality level = MQ100

1.09631.0100 1.09631.0500
1.09631.0100EA 166 EUR
1.09631.0100 1.09631.0500
Charcoal activated 0.3-0.5 mm (35-50 mesh) for gas chromatography, Supelco®
Charcoal activated
Formula: C
MW: 12,01 g/mol
Melting Pt: 3550 °C
Density: 2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now

Specification Test Results

Resolution 7.0 - 11.0
Bulk density 0.4 - 0.5 g/cm³
Zn (Zinc) ≤ 0.002 %
Particle size (0.3 - 0.5 mm) ≥ 70.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR