Airborne microorganism samplers, MAS-100 CG Ex®
Supplier: Merck
1.09327.0001EA 0 EUR
1.09327.0001
Airborne microorganism samplers, MAS-100 CG Ex®
Samplers Air Samplers
Compressed gases must also be tested routinely for microbial contamination.
  • Programmable delay and individually adjustable pre-selected volumes
  • Sampling head tested by an accredited institute
  • Pre-programmed for compressed CO₂, N₂ or air
  • Programmable for many more gas types
  • Connection for PC or external adapter
  • Standard 90 mm petri dishes can be used - The sampling on standard petri dishes is performed under pressure
The different types of gases used in the pharmaceutical or food industry are recognised by the instrument and compensated automatically for their individual physical characteristics.
Certifications: ISO 14698-1/2
Delivery information: Supplied in a trolley case.