Would you like to log in again?
Adhesive PCR film and foil seals
Supplier: Eppendorf
Masterclear®
Effective adhesive sealing prevents evaporation loss during PCR.
  • Removal from the plate with no residue
  • Autoclavable (121 °C, 20 min)
  • PCR Film: Sample can be visually monitored through the transparent film
  • PCR Foil: Easily pierced; no sticking of the pipette tip (ideal for automated systems)
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Self-adhesive PCR sealing film, PCR clean 100 713-0005
Self-adhesive PCR sealing foil, PCR clean 100 713-0006
Masterclear® real-time PCR film, self-adhesive, PCR clean 100 713-0019
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty