Microplate readers, UV/Vis absorbance, Biochrom EZ Read 2000
736-0863EA 15540 EUR
WAPR80-4002-20
Microplate readers, UV/Vis absorbance, Biochrom EZ Read 2000
The Biochrom EZ Read 2000 is a PC-controlled microplate reader, which supports multiple plate formats and is capable of measuring at any wavelength in 1 nm increments between 340 and 800 nm. The monochromator-based system allows wave scanning and multiple measurements at different wavelengths in the same well - important for contamination detection, sample purity analysis, optimal peak absorbance detection and product formation studies. Automatic calibration ensures accurate readings.
Applications include ELISAs, enzyme immunoassays, total protein quantitation (BCA, Lowry, Bradford), cell proliferation, and cell survival assays, plus cell based assays where shaking is required to keep cells in suspension, enzyme kinetic assays, assays requiring wave scans or spectral scans, and assays measuring the changes in absorbance of NADH at 340 nm.
  • Multiple plate formats: 12-, 24-, 48- and 96-well plates
  • Flexible wavelength selection: Ideal for a variety of assays
  • Single or dual wavelength measurement
  • Timed interval reading: Ideal for kinetic assays
  • Linear shaking in four modes
  • Data analysis of endpoint measurements with Galapagos software package
Applications include ELISAs, enzyme immunoassays, total protein quantitation (BCA, Lowry, Bradford), cell proliferation, and cell survival assays, plus cell based assays where shaking is required to keep cells in suspension, enzyme kinetic assays, assays requiring wave scans or spectral scans, and assays measuring the changes in absorbance of NADH at 340 nm.
The readers come with Galapagos PC software for instrument control and data analysis. Featuring ‘click and drag’ functionality, Galapagos software enables configuration of plate layouts, multiple assays per plate and multiple plates per analysis.
Ordering information: For full details of options and accessories including QC plate for instrument validation and spare lamps, please contact your local Avantor sales office.
Delivery information: Supplied with Galapagos PC software (compatible with Windows® 7 and 8, and XP onwards), manual, power cord and USB cable.