3-(3-Fluorophenylcarbamoyl)phenylboronic acid ≥98%
|
Warning
|
Synonyms:
3-(3-Fluorophenylcarbamoyl)benzeneboronic acid
H52959.03EA
163
EUR
H52959.03
3-(3-Fluorophenylcarbamoyl)phenylboronic acid ≥98%
3-(3-Fluorophenylcarbamoyl)phenylboronic acid
Formula:
C₁₃H₁₁BFNO₃ MW: 259,04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09027207 CAS Number: 874288-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...