4-(3-Fluorophenylcarbamoyl)phenylboronic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-(3-Fluorophenylcarbamoyl)benzeneboronic acid
, (4-((3-Fluorophenyl)carbamoyl)phenyl)boronic acid
H53192.MDEA
0
EUR
H53192.MD
4-(3-Fluorophenylcarbamoyl)phenylboronic acid ≥98%
4-(3-Fluorophenylcarbamoyl)phenylboronic acid
Formula:
C₁₃H₁₁BFNO₃ MW: 259,04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09027204 CAS Number: 874288-05-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...