1,1'-Bis(dicyclohexylphosphino)ferrocene ≥98%
Supplier: Thermo Scientific
H27388.MCEA
55.2
EUR
H27388.MC
H27388.03
1,1'-Bis(dicyclohexylphosphino)ferrocene ≥98%
1,1'-Bis(dicyclohexylphosphino)ferrocene
Formula:
C₃₄H₅₂FeP₂ MW: 570,5057 g/mol Melting Pt: 134…136 °C |
MDL Number:
MFCD09038820 CAS Number: 146960-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...