2,3,6-Trimethylquinoxaline ≥97%
Supplier: Thermo Scientific
|
Warning
|
B21432.06EA
108
EUR
B21432.06
B21432.03
2,3,6-Trimethylquinoxaline ≥97%
2,3,6-Trimethylquinoxaline
Formula:
C₁₁H₁₂N₂ MW: 172,23 g/mol Melting Pt: 91…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047593 CAS Number: 17635-21-1 EINECS: 241-618-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...