Okadaic acid potassium salt

Supplier: Enzo Life Sciences

Synonyms: 9,10-deepithio-9,10-didehydroacanthifolicin potassium saltHalochondrine A potassium saltOA potassium salt

ALEXALX350063C050EA 180 EUR
ALEXALX350063C050 ALEXALX350063M001 ALEXALX350063C100
Okadaic acid potassium salt
Okadaic acid potassium salt
Formula: C₄₄H₆₇O₁₃·K
MW: 843,11 g/mol
MDL Number: MFCD00214356
CAS Number: 155751-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR