Ethyl-2-chloro-4-oxazolecarboxylate 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Ethyl 2-chlorooxazole-4-carboxylate
H31521.03EA
210
EUR
H31521.03
Ethyl-2-chloro-4-oxazolecarboxylate 95%
Ethyl 2-chloro-4-oxazolecarboxylate
Formula:
C₆H₆ClNO₃ MW: 175,57 g/mol Melting Pt: 97…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD06660120 CAS Number: 460081-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...