5-(3-Bromophenyl)oxazole ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 5-(3-Bromophenyl)-1,3-oxazole

H34161.09EA 310 EUR
H34161.09 H34161.03
5-(3-Bromophenyl)oxazole ≥97%
5-(3-Bromophenyl)oxazole
Formula: C₉H₆BrNO
MW: 224,06
Melting Pt: 50…52 °C
Storage Temperature: Ambient
MDL Number: MFCD05668970
CAS Number: 243455-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR