5-(4-Fluorophenyl)oxazole

Supplier: Apollo Scientific

APOSPC4174C-1GEA 71.4 EUR
APOSPC4174C-1G
5-(4-Fluorophenyl)oxazole
5-(4-Fluorophenyl)oxazole
Formula: C₉H₆FNO
MDL Number: MFCD00085087
CAS Number: 128101-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR