3-(1H-Tetrazol-5-ylcarbamoyl)phenylboronic acid hydrochloride ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 3-(1H-Tetrazol-5-ylcarbamoyl)benzeneboronic acid hydrochloride

H52537.MDEA 29.8 EUR
H52537.MD H52537.03
3-(1H-Tetrazol-5-ylcarbamoyl)phenylboronic acid hydrochloride ≥97%
3-(1H-Tetrazol-5-ylcarbamoyl)phenylboronic acid hydrochloride
Formula: C₈H₉BClN₅O₃
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD06659888
CAS Number: 850567-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR