N-(2-Bromo-4,6-dichlorophenyl)benzenesulphonamide ≥97%

Supplier: Thermo Scientific
Warning

H56293.MDEA 140 EUR
H56293.MD H56293.03
N-(2-Bromo-4,6-dichlorophenyl)benzenesulphonamide ≥97%
N-(2-Bromo-4,6-dichlorophenyl)benzenesulphonamide
Formula: C₁₂H₈BrCl₂NO₂S
Storage Temperature: Ambient
MDL Number: MFCD22682861
CAS Number: 1421262-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR