4-Bromo-N-(3-(trifluoromethyl)phenyl)benzamide ≥97%

Supplier: Thermo Scientific
Warning

H58046.MCEA 75.7 EUR
H58046.MC H58046.ME
4-Bromo-N-(3-(trifluoromethyl)phenyl)benzamide ≥97%
4-Bromo-N-(3-(trifluoromethyl)phenyl)benzamide
Formula: C₁₄H₉BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD00421306
CAS Number: 56661-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR