N-(2-Bromo-5-nitrophenyl)benzenesulphonamide ≥97%

Supplier: Thermo Scientific
Warning

H56210.03EA 448 EUR
H56210.03 H56210.MD
N-(2-Bromo-5-nitrophenyl)benzenesulphonamide ≥97%
N-(2-Bromo-5-nitrophenyl)benzenesulphonamide
Formula: C₁₂H₉BrN₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD22682850
CAS Number: 1421261-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR