4-Chlor-3-(isopropylcarbamoyl)phenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Chloro-3-(isopropylcarbamoyl)benzeneboronic acid(4-Chloro-3-(isopropylcarbamoyl)phenyl)boronic acid

H52679.MDEA 110 EUR
H52679.MD
4-Chlor-3-(isopropylcarbamoyl)phenylboronic acid ≥98%
4-Chlor-3-(isopropylcarbamoyl)phenylboronic acid
Formula: C₁₀H₁₃BClNO₃
MW: 241,48 g/mol
Melting Pt: 254…260 °C
Storage Temperature: Ambient
MDL Number: MFCD07783866
CAS Number: 871332-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR