2-Chloro-6-fluoro-3-methoxybenzaldehyde ≥97%

Supplier: Thermo Scientific
Warning

H26403.03EA 73.2 EUR
H26403.03 H26403.09
2-Chloro-6-fluoro-3-methoxybenzaldehyde ≥97%
2-Chloro-6-fluoro-3-methoxybenzaldehyde
Formula: C₈H₆ClFO₂
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD04115941
CAS Number: 149949-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR