4-Chloro-2-fluoro-3-methoxybenzaldehyde ≥97%

Warning

H33274.03EA 115 EUR
H33274.03 H33274.06
4-Chloro-2-fluoro-3-methoxybenzaldehyde ≥97%
4-Chloro-2-fluoro-3-methoxybenzaldehyde
Formula: C₈H₆ClFO₂
MW: 188,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687172
CAS Number: 1002344-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR