4-Chloro-3-ethoxy-2-fluorobenzaldehyde ≥97%

Supplier: Thermo Scientific
Warning

H33381.06EA 353 EUR
H33381.06 H33381.03
4-Chloro-3-ethoxy-2-fluorobenzaldehyde ≥97%
4-Chloro-3-ethoxy-2-fluorobenzaldehyde
Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD19687180
CAS Number: 1323966-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR