Sulphachloropyridazine, Supelco®
Supplier: Merck
Synonyms:
4-Amino-N-(6-chloropyridazin-3-yl)benzenesulphonamide
M-Clarity™ quality level = MQ200
46778-250MGEA
60.9
EUR
46778-250MG
Sulphachloropyridazine, Supelco®
Sulphachloropyridazine
Formula:
C₁₀H₉ClN₄O₂S MW: 284,7221 g/mol |
MDL Number:
MFCD00057371 CAS Number: 80-32-0 EINECS: 201-269-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...