4-Fluoro-3-(methoxy(methyl)carbamoyl)phenylboronic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Fluoro-3-[methoxy(methyl)carbamoyl]benzeneboronic acid
, (4-Fluoro-3-(methoxy(methyl)carbamoyl)phenyl)boronic acid
H52512.MDEA
0
EUR
H52512.MD
4-Fluoro-3-(methoxy(methyl)carbamoyl)phenylboronic acid ≥98%
4-Fluoro-3-(methoxy(methyl)carbamoyl)phenylboronic acid
Formula:
C₉H₁₁BFNO₄ MW: 227,00 g/mol Melting Pt: 160…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD08235106 CAS Number: 874290-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...