4-Fluoro-3-(2-hydroxyethylcarbamoyl)phenylboronic acid ≥98%
|
Warning
|
Synonyms:
4-Fluoro-3-(2-hydroxyethylcarbamoyl)benzeneboronic acid
H52934.03EA
341
EUR
H52934.03
4-Fluoro-3-(2-hydroxyethylcarbamoyl)phenylboronic acid ≥98%
4-Fluoro-3-(2-hydroxyethylcarbamoyl)phenylboronic acid
Formula:
C₉H₁₁BFNO₄ MW: 227,00 g/mol Melting Pt: 89…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD08235044 CAS Number: 874219-25-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...