4-Fluoro-3-(methoxycarbamoyl)phenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Fluoro-3-(methoxycarbamoyl)benzeneboronic acid

H52875.MDEA 0 EUR
H52875.MD
4-Fluoro-3-(methoxycarbamoyl)phenylboronic acid ≥98%
4-Fluoro-3-(methoxycarbamoyl)phenylboronic acid
Formula: C₈H₉BFNO₄
MW: 212,97 g/mol
Melting Pt: 213…215 °C
Storage Temperature: Ambient
MDL Number: MFCD08235105
CAS Number: 913835-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR