N-(4-Bromophenyl)-3-(trifluoromethyl)benzamide ≥97%
Supplier: Thermo Scientific
|
Warning
|
H57015.MDEA
102
EUR
H57015.MD
H57015.03
N-(4-Bromophenyl)-3-(trifluoromethyl)benzamide ≥97%
N-(4-Bromophenyl)-3-(trifluoromethyl)benzamide
Formula:
C₁₄H₉BrF₃NO MW: 344,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03937564 CAS Number: 562080-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...