3-Chloro-4-methylbenzenesulphonamide ≥97%
|
Warning
|
H32513.09EA
204
EUR
H32513.09
3-Chloro-4-methylbenzenesulphonamide ≥97%
3-Chloro-4-methylbenzenesulphonamide
Formula:
C₇H₈ClNO₂S MW: 205,66 g/mol Melting Pt: 135…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025381 CAS Number: 51896-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...