8-Methoxy-5-quinolineboronic acid pinacol ester 96%
Supplier: Thermo Scientific
H54445.03EA
324
EUR
H54445.03
H54445.MD
8-Methoxy-5-quinolineboronic acid pinacol ester 96%
8-Methoxy-5-quinolineboronic acid pinacol ester
Formula:
C₁₆H₂₀BNO₃ MW: 285,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20527214 CAS Number: 916162-54-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...