2-(Trimethylsilyl)pyridine ≥97%

Supplier: Thermo Scientific
Warning

H26885.03EA 39.3 EUR
H26885.03 H26885.09
2-(Trimethylsilyl)pyridine ≥97%
2-(Trimethylsilyl)pyridine
Formula: C₈H₁₃NSi
MW: 151,28
Boiling Pt: 47…49 °C (5 mmHg)
Density: 0,911
Flash Pt: 52 °C (126 °F)
Storage Temperature: Ambient
MDL Number: MFCD06797181
CAS Number: 13737-04-7
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR