3-Phenoxythiophene ≥97%

Supplier: Thermo Scientific
Danger

B25288.03EA 41.1 EUR
B25288.03 B25288.14 B25288.06
3-Phenoxythiophene ≥97%
3-Phenoxythiophene
Formula: C₁₀H₈OS
Storage Temperature: Ambient
MDL Number: MFCD00178747
CAS Number: 63285-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR