2,4,6-Trimethyl-4-heptanol ≥97%

Supplier: Thermo Scientific
Warning

B20059.06EA 72.3 EUR
B20059.06 B20059.03 B20059.14
2,4,6-Trimethyl-4-heptanol ≥97%
2,4,6-Trimethyl-4-heptanol
Formula: C₁₀H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD00048354
CAS Number: 60836-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR