2-Fluoro-4-methoxybenzamide ≥97%

Supplier: Thermo Scientific
Warning

H31636.03EA 69.3 EUR
H31636.03 H31636.06
2-Fluoro-4-methoxybenzamide ≥97%
2-Fluoro-4-methoxybenzamide
Formula: C₈H₈FNO₂
Melting Pt: 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD01117440
CAS Number: 874804-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR